missing translation for 'onlineSavingsMsg'
Learn More

Water, B&J Brand™ LC-MS, for LC-MS and HPLC, Honeywell Burdick & Jackson

Supplier:  Honeywell-Burdick and Jackson LC36525

 View more versions of this product

Catalog No. 6003077


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Water
7732-18-5
100.0000%
2050024
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18
Glass Bottle, Gl45
4 x 2.5 L
B&J Brand™ LC-MS, for LC-MS and HPLC
100°C
H2O
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
LC-MS
0.9971 g/cm3

Chemical Identifiers

7732-18-5
18.015
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

SDS
Documents

Documents

Promotions

Promotions