missing translation for 'onlineSavingsMsg'
Learn More

Water, B&J Brand™ LC-MS, for LC-MS and HPLC, Honeywell Burdick & Jackson

Catalog No. p-7112035
Change view
Click to view available options
Quantity:
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Glass Bottle, Gl45
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6003076 6 x 1 L Glass Bottle, Gl45
6003077 4 x 2.5 L Glass Bottle, Gl45
6003078 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6003076 Supplier Honeywell Chemicals Supplier No. LC3651
Only null left

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Boiling Point 100°C
CAS Max % 100.0000%
Beilstein 2050024
Formula Weight 18
Grade LC-MS
Packaging Glass Bottle, Gl45
Density 0.9971 g/cm3
Quantity 6 x 1 L
Chemical Name or Material Water, B&J Brand™ LC-MS, for LC-MS and HPLC
ShelfLife 547 days from date of manufacture