missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, for LC-MS and HPLC, Honeywell Burdick & Jackson
Specifications
Specifications
| Boiling Point | 100°C |
| CAS Max % | 99.9000%, 0.1000% |
| Density | 0.9982 g/cm3 |
| Grade | HPLC |
| Packaging | Glass Bottle, Gl45 |
| Quantity | 6 x 1 L |
| Chemical Name or Material | Water with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, for LC-MS and HPLC |
Safety and Handling
| ShelfLife | 365 days from date of manufacture |