missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Waste Disposal Labels - Adhesive Vinyl

Use these durable labels to easily identify and sort waste for disposal.

Supplier:  Brady™ 121433

 View more versions of this product

Catalog No. 19104933


Only null left

Description

Description

  • All HazMat Shipping Labels meet D.O.T. Requirements HM-181 Regulation.
  • Permanent acrylic adhesive sticks to a wide variety of surfaces
  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
  • Up to 3 year average outdoor durability
Specifications

Chemical Identifiers

Not Autoclavable
Permanent Acrylic, Self-adhesive
Vinyl
Label
100/Pk.
6 x 6 in.
6 in.
Label
Label
Single-side Printable
English
6 x 6 in.
6 in.

Specifications

Not Autoclavable
Label
Black on Yellow
Vinyl
Label
English
Rectangle
6 x 6 in.
160.02 mm
Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200
Permanent Acrylic, Self-adhesive
Label
Single-side Printable
USED ABSORBENTS
100/Pk.
6 x 6 in.
6 in.
6 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions