missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Buffer B for Dynabeads™ mRNA Purification Kits

Catalog No. 11900D
Change view
Click to view available options
Quantity:
1 x 60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11900D 1 x 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11900D Supplier Invitrogen™ Supplier No. 11900D

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Wash Buffer B is included in the following Dynabeads™ mRNA purification kits.

This Wash Buffer B is included in the following Dynabeads™ mRNA purification kits: Cat. Nos. 61006, 61011, 61012, and 61021. It is made available separately for applications that require more Wash Buffer B than is provided in the kit.

Specifications

Content And Storage 1 x 60 mL, store at 2°C to 8°C
Packaging Type Kit
Product Type Wash Buffer B
Product Line Dynabeads
Quantity 1 x 60 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.