missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hughes Wall Mounted Combination Shower
Hughes Wall Mounted Combination Shower provides initial on-site treatment to eye/face or full body exposure to hazards
Specifications
| Activation | Shower: Hand Lever, Eye and Face Wash: Pull Lid Forward |
|---|---|
| Bowl Size (Metric) | 241 mm |
| Certifications/Compliance | ANSI Z358.1-2014 and EN15154 Compliant |
| Depth (English) | 16 in. |
| Depth (Metric) | 406 mm |
Description
Features:
- Specially designed valve that includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure
- Low operating torque offers reliable, easy on/off operation
- Durable, coated stainless steel pull handle with nylon valve connection is easy to grip and activate for immediate water drenching.
- Designed for high flow rates that meet or exceed ANSI standards, the nozzle offers optimal spray distribution for user protection
- Unlike shower rose designs, this compact nozzle reduces the potential for insect nesting and eliminates high replacement costs associated with cracked or dented plastic spray heads
- Made of durable, corrosion-resistant acrylic capped ABS, the eye/face wash with high, ANSI-compliant flow rate features an integral lid connected to the water valve for automatic operation—activate by simply pulling down the lid.
- Easy-to-reach flow regulator valve positioned on the side of the bowl adjusts to suit the severity of the accident.
- Solid construction for added corrosion resistance. Includes an inspection record tag for recording service and test history
- Covered bowl ensures eyewash nozzles and bowl remain clean and free from contamination
- Twin aerated diffusers provide a soft, scrubbing action to the eyes and face
- Removable, self-draining nozzle constructed from durable nylon
- High-performance quarter-turn ball valve made of 316L stainless steel endure the harshest environments and site conditions
- Includes a universal eye wash and safety shower sign for enhanced visibility
- Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements
Compliance
ANSI Z358.1-2014 and EN15154 Compliant
Warranty
1 Year
Specifications
| Shower: Hand Lever, Eye and Face Wash: Pull Lid Forward | |
| ANSI Z358.1-2014 and EN15154 Compliant | |
| 406 mm | |
| Emergency Shower and Eye Wash with Water | |
| 406 mm | |
| 1.25 in. (F) NPT | |
| Wall | |
| Safety Shower and Emergency Eye Wash | |
| 1219 mm |
| 241 mm | |
| 16 in. | |
| 3 gpm | |
| 16 in. | |
| 1 Unit | |
| Nylon 6, Chrome-plated Brass, Stainless Steel Mesh | |
| 29 to 87 psi | |
| 48 in. |