missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ W 54011

Catalog No. 545550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
545550 50 mg
545510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 545550 Supplier Tocris Bioscience™ Supplier No. 5455/50
Only null left

High affinity C5a receptor antagonist

W 54011 is a high affinity C5a receptor antagonist (Ki = 2.2 nM). Inhibits C5a-induced generation of reactive superoxide species, chemotaxis and intracellular Ca2+ mobilization in human neutrophils (IC50 values are 1.6, 2.7 and 3.1 nM, respectively).

Specifications

Chemical Name or Material N-[[4-(Dimethylamino)phenyl]methyl]-1,2,3,4-tetrahydro-7-methoxy-N-[4-(1-methylethyl)phenyl]-1-naphthalenecarboxamide hydrochloride
CAS 405098-33-1
Quantity 50 mg
Target Complement Signaling inhibitors
Molecular Formula C30H36N2O2.HCl
Formula Weight 493.08
Purity >98%