missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ W 54011
Description
W 54011 is a high affinity C5a receptor antagonist (Ki = 2.2 nM). Inhibits C5a-induced generation of reactive superoxide species, chemotaxis and intracellular Ca2+ mobilization in human neutrophils (IC50 values are 1.6, 2.7 and 3.1 nM, respectively).
Specifications
Specifications
| Chemical Name or Material | N-[[4-(Dimethylamino)phenyl]methyl]-1,2,3,4-tetrahydro-7-methoxy-N-[4-(1-methylethyl)phenyl]-1-naphthalenecarboxamide hydrochloride |
| CAS | 405098-33-1 |
| Quantity | 10 mg |
| Target | Complement Signaling inhibitors |
| Molecular Formula | C30H36N2O2.HCl |
| Formula Weight | 493.08 |
| Purity | >98% |