missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ VUF 11222
Description
VUF 11222 is a high affinity non-peptide CXCR3 agonist (pKi = 7.2).
Specifications
Specifications
| Chemical Name or Material | 2′-Bromo-N-[[(1R,5S)-6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl]methyl]-N,N-dimethyl-[1,1′-biphenyl]-4-methanaminium iodide |
| CAS | 1414376-84-3 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Chemokine Receptor Agonists |
| Molecular Formula | C25H31BrIN |
| Formula Weight | 552.33 |
| Purity | >98% |