missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ VOST Stack Sampling Tube

Catalog No. 111001617
Change view
Click to view available options
Material:
Glass, Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
111001617 Glass, Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001617 Supplier MilliporeSigma™ Supelco™ Supplier No. 20074U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to meet the specifications in US EPA SW-846, Method 0030

  • Individually numbered tubes
  • Sealed with a stainless steel Swagelok fitting
  • Placed in a glass storage container
  • Tested for background and back pressure
  • Pre-conditioned

Specifications

Material Glass, Stainless Steel
Includes 10 Tubes, 10 Tube Adapters, 10 Male Luer ⅛ Fittings
Packaging Quantity 1/Kit
For Use With (Application) Air monitoring, Environmental, Industrial hygiene
Matrix Tenax™ TA Porous Polymer
Description Particle Size: 35-60 Mesh
Methods US EPA SW-846, Method 0030
Type Volatile Organic Sampling Train (VOST) Tube