missing translation for 'onlineSavingsMsg'
Learn More

Bulbtronics™ Voltarc™ Germicidal Lamp

Specifications

Base Style Single Pin Slimline (FA8)
Light Source Germicidal UV-C
Lighting Life 7500 hr.
Wavelength 254 nm
Amperage 400 mA
View More Specs

Products 2
Catalog Number Mfr. No. Wattage Price Quantity & Availability  
Catalog Number Mfr. No. Wattage Price Quantity & Availability  
14395339
Encompass
View Documents Promotion Details
Bulbtronics™
BBT171
39 W N/A
Only null left
 
14395345
Encompass
View Documents Promotion Details
Bulbtronics™
BBT187
65 W N/A
Only null left
 
Description

Description

Specifications

Specifications

Single Pin Slimline (FA8)
7500 hr.
400 mA
0.62 in.
T5
Germicidal UV-C
254 nm
Rapid Start
1.58 cm
Linear Double-ended Lamp
Videos
SDS
Documents

Documents