missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ Turbo 4 Ultrafiltration Unit

Catalog No. 14558714
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
5 kDa
50 kDa
Quantity:
100/Pk.
25/Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
14558714 100 kDa 100/Pk.
14558703 3 kDa 25/Pk.
14558704 3 kDa 100/Pk.
14558705 5 kDa 25/Pk.
14558706 5 kDa 100/Pk.
14558707 10 kDa 25/Pk.
14558708 10 kDa 100/Pk.
14558709 30 kDa 25/Pk.
14558710 30 kDa 100/Pk.
14558711 50 kDa 25/Pk.
14558712 50 kDa 100/Pk.
14558713 100 kDa 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14558714 Supplier Sartorius Supplier No. VS04T42

Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.

  • High flux PES membrane
  • Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
  • Universal rotor compatibility
  • Volume graduation for easy detection / monitoring of the concentrate volume
  • UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket

Specifications

Description Turbo 4 Ultrafiltration Unit
Disposable Single-use
Filter Type Ultrafiltration
Material (Membrane) PES
MWCO 100 kDa
Sterility Non-sterile
For Use With (Application) Concentration, Desalting, buffer exchange
Max. Force Swing-out rotor : 4000 x gAngle rotor : 7500 x g
Quantity 100/Pk.
Sample Type Sample type: Nanoparticles; Peptide; Protein
Type Centrifugal Concentrator
Volume (Metric) 4 mL
Length (Metric) 1225 mm
For Use With (Equipment) Centrifuge
Sample Volume 4 mL
Show More Show Less