Learn More
Sartorius Vivaspin™ Turbo 4 Ultrafiltration Unit
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
Supplier: Sartorius VS04T11
Description
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
- High flux PES membrane
- Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
- Universal rotor compatibility
- Volume graduation for easy detection / monitoring of the concentrate volume
- UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket
Specifications
| Turbo 4 Ultrafiltration Unit | |
| Ultrafiltration | |
| PES | |
| 4 mL | |
| Concentration, Desalting, buffer exchange | |
| 25/Pk. | |
| Centrifugal Concentrator | |
| 1225 mm |
| Single-use | |
| Centrifuge | |
| 5 kDa | |
| Non-sterile | |
| Swing-out rotor : 4000 x gAngle rotor : 7500 x g | |
| Sample type: Nanoparticles; Peptide; Protein | |
| 2 to 4 mL |