missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaspin Turbo 15RC
Description
- Fastest sample concentration with maximum target molecule recoveries
- Simplest, pipette-friendly retentate retrieval
- Highest chemical and temperature resistance
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Format | Spin Column |
| For Use With (Equipment) | Centrifuge |
| Material | SBC |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 10 kDa |
| Sample Volume | 4 to 15 mL |
| Sterility | Non-sterile |
| Quantity | 12 Pk. |
| Type | Centrifugal |
| Show More |