missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivapore 5
Description
- Concentrate Equipment-Free: Solvent absorption is ideal where centrifuges are not available and for high-throughput applications.
- Fill and Go: Simply add your sample and leave it to reach the desired final volume, with no risk of concentration to dryness.
- Tailored Process Monitoring: Printed markings help to reduce the guesswork when each sample needs to reach its own specific concentration factor.
- Maintained Sample Integrity: Self-contained, single use units ensure contamination-free concentration of individual samples.
- Sensitive Molecule Friendly: Benchtop or refrigerated low pressure processing for shear and temperature sensitive molecules.
- Expand or Exchange: Optional reservoirs provide the flexibility to double sample capacity or concentrate and desalt in one step
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Material (Membrane) | PES |
| MWCO | 7.5 kDa |
| Sterility | Non-sterile |
| Type | Solvent Absorption |
| Volume (Metric) | 5 mL |
| Sample Volume | 5 mL |