missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaflow™ 200 Crossflow Cassettes
Offers a standard for ease of use, reliability and flexibility unrivalled by other filtration systems. Sartorius Vivaflow 200 Crossflow Cassettes are ready to use, reusable laboratory crossflow cassettes for concentrating up to 5L samples. Available in different MWCOs and membrane types for different application needs.
$826.31 - $826.31
Specifications
| Filter Type | Ultrafiltration |
|---|---|
| Format | Cassette |
| For Use With (Equipment) | Labscale TFF System |
| Material | Acrylic |
| Sample Volume | Up to 2.5 L |
| Catalog Number | Mfr. No. | Material (Membrane) | MWCO | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material (Membrane) | MWCO | Price | Quantity | ||||
|
14-558-320
|
Sartorius
VF20P2 |
PES | 30 kDa |
Each for $826.31
|
|
||||
|
14-558-318
|
Sartorius
VF20P0 |
PES | 10 kDa |
Each for $826.31
|
|
||||
|
14-558-322
|
Sartorius
VF20P4 |
PES | 100 kDa |
Each for $826.31
|
|
||||
|
14-558-315
|
Sartorius
VF20H0 |
Cellulose | 10 kDa |
Each for $826.31
|
|
||||
|
14-558-321
|
Sartorius
VF20P3 |
PES | 50 kDa |
Each for $826.31
|
|
||||
|
14-558-317
|
Sartorius
VF20H2 |
Cellulose | 30 kDa |
Each for $826.31
|
|
||||
|
14-558-319
|
Sartorius
VF20P1 |
PES | 5 kDa |
Each for $826.31
|
|
||||
|
14-558-323
|
Sartorius
VF20P7 |
PES | 0.2 μm |
Each for $826.31
|
|
||||
|
14-558-314
|
Sartorius
VF20H4 |
Cellulose | 100 kDa |
Each for $826.31
|
|
||||
|
14-558-316
|
Sartorius
VF20H1 |
Cellulose | 5 kDa |
Each for $826.31
|
|
||||
Description
Rapid, Efficient Concentration- Optimally suited for cell culture supernatant concentration, virus concentration and water concentration
- Utilizes a 200cm2 PES or Hydrosart membrane for high flow rate
- Thin-channel, flip-flow recirculation path provides high crossflow velocities with minimum pump speed
- Fast concentration using one or two cassettes
- A single module typically concentrates 1L by a factor of 50 in less than 30 minutes
- Two modules used in parallel concentrates 5L by a factor of 50 in under 75 minutes
- Near total recovery of the concentrate is achieved with a single rinse
- A 500ml sealed diafiltration reservoir can be added for rapid and convenient buffer exchange
- One unit comes with all necessary accessories (except pump and pump head)
- Includes tubing, pressure indicator, flow restrictor and high pressure pump tubing
- Required equipment: peristaltic pump with a size 16 pump head
- Can be easily cleaned and re-used several times
- Clear housing for process monitoring
- Ready-to-use, disposable cassettes
- Easily set up two cassettes to run simultaneously for faster concentration
- Use a pre-assembled Y connector with the correct tubing sizes and fittings
- Size 16 tubing is used for connecting the two Vivaflow 200 units
- Size 15 tubing to connect to the pump, with a size 15 pump head
Specifications
| Ultrafiltration | |
| Labscale TFF System | |
| Up to 2.5 L | |
| 200 cm2 | |
| Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings | |
| 200 to 400mL/min. | |
| 140°F | |
| Crossflow Cassette | |
| 5.3 mL | |
| 12.6 cm | |
| 1 cm | |
| 0.04 cm |
| Cassette | |
| Acrylic | |
| Non-sterile | |
| Cell culture supernatant concentration, virus concentration and water concentration | |
| 60psi | |
| 1 Each | |
| 60°C | |
| 5 L | |
| 4.96 x 1.49 x 5.43 in. (12.6 x 3.8 x 13.8cm) | |
| 3.8 cm | |
| 13.8 cm |