missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Viva™ HPLC Columns with C18 Column Packing, 5μm Particle Size

Catalog No. p-3216645
Encompass
Change view
Click to view available options
Length (Metric):
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
4.6 mm
Catalog No. Length (Metric) Diameter (Metric)
06739207 100 mm 4.6 mm
06739227 250 mm 4.6 mm
06739221 150 mm 2.1 mm
3 options

Catalog No. 06739207

Supplier: Restek™ RK 9514515

Only null left

Wide bore silica columns excellent for general-purpose applications and separating peptides or proteins

  • Highly base-deactivated wide pore packing exhibits excellent peak shape
  • Rugged, spherical particles
  • 300Å pore size with a narrow pore size distribution—high proportion of pore/surface area available for large molecule and biomolecule separations
  • Carbon load: 9%
  • pH range: 2.5 to 10 pH range
  • Temperature limit: 80°C

Specifications

Diameter (Metric) 4.6 mm
Film Thickness 5 μm
Flow Rate 1 mL/min
For Use With Analyzing Large Molecules and Biomolecules
Length (Metric) 100 mm
Ligand Type Monomeric C18
Max. Pressure 4000 psi (275 bar)
Particle Shape Spherical
Particle Size 5 μm
pH 2.5 to 10
Pore Size 300 Å
Temperature 80°C
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Type Column
Show More Show Less