Learn More
Thermo Scientific Chemicals Vitamin D3, 99%
Vitamin D3, CAS # 67-97-0, also known as cholecalciferol, is a steroid hormone produced in the skin or obtained from dietary sources. It is an essential mediator of bone calcium metabolism. | CAS: 67-97-0 | C27H44O | 384.648 g/mol
Supplier: Thermo Scientific Chemicals B2252406
Description
Vitamin D3 is used as an essential mediator of intestinal calcium absorption and bone calcium metabolism. It is also used as a therapeutic vitamin. Further, it is used as a nutritional supplement and to fortify foods. In addition to this, it is used as pharmaceutical drug and used in the treatment of respiratory diseases involving chronic inflammation such as asthma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Vitamin D3 is a steroid hormone generated in the skin when the 7-dehydrocholesterol interacts with ultraviolet irradiation. It can also be found in several types of food for daily intake.
- Vitamin D3 binds to vitamin D receptors, thus modulating gene expression
Applications
- It presents in vitro antiproliferative and antimetastatic activities on breast, colon, and prostate cancer cells
Specifications
| Vitamin D3 | |
| 83°C to 87°C | |
| C27H44O | |
| 5 g | |
| 2339331 | |
| vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum | |
| QYSXJUFSXHHAJI-YRZJJWOYSA-N | |
| (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol | |
| 5280795 | |
| 384.65 | |
| Crystalline Powder |
| 67-97-0 | |
| Glass bottle | |
| MFCD00078131 | |
| UN2811 | |
| Air and light sensitive | |
| Soluble in ethanol and methanol. Slightly soluble in vegetable oils. Insoluble in water. | |
| CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C | |
| 384.648 | |
| CHEBI:28940 | |
| 99% |
Chemical Identifiers
| 67-97-0 | |
| 384.648 | |
| QYSXJUFSXHHAJI-YRZJJWOYSA-N | |
| 5280795 | |
| (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol |
| C27H44O | |
| MFCD00078131 | |
| vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum | |
| CHEBI:28940 | |
| CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C |
Safety and Handling
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H372
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 200-673-2
RTECSNumber : VS2900000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only