missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vitamin D3, 99%

Catalog No. p-7076517
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2252403 1 g
AAB2252406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2252403 Supplier Thermo Scientific Chemicals Supplier No. B2252403
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Vitamin D3, CAS # 67-97-0, also known as cholecalciferol, is a steroid hormone produced in the skin or obtained from dietary sources. It is an essential mediator of bone calcium metabolism. | CAS: 67-97-0 | C27H44O | 384.648 g/mol

Vitamin D3 is used as an essential mediator of intestinal calcium absorption and bone calcium metabolism. It is also used as a therapeutic vitamin. Further, it is used as a nutritional supplement and to fortify foods. In addition to this, it is used as pharmaceutical drug and used in the treatment of respiratory diseases involving chronic inflammation such as asthma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vitamin D3 is a steroid hormone generated in the skin when the 7-dehydrocholesterol interacts with ultraviolet irradiation. It can also be found in several types of food for daily intake.
  • Vitamin D3 binds to vitamin D receptors, thus modulating gene expression

Applications

  • It presents in vitro antiproliferative and antimetastatic activities on breast, colon, and prostate cancer cells

Chemical Identifiers

CAS 67-97-0
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.648
MDL Number MFCD00078131
InChI Key QYSXJUFSXHHAJI-YRZJJWOYSA-N
Synonym vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum
PubChem CID 5280795
ChEBI CHEBI:28940
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C

Specifications

Melting Point 83°C to 87°C
Packaging Glass bottle
Quantity 1 g
UN Number UN2811
Beilstein 2339331
Sensitivity Air and light sensitive
Solubility Information Soluble in ethanol and methanol. Slightly soluble in vegetable oils. Insoluble in water.
Formula Weight 384.65
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Vitamin D3
Show More Show Less
Hazard Category H300+H310+H330-H372
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 200-673-2
RTECSNumber VS2900000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only