accessibility menu, dialog, popup

UserName

CAS RN 67-97-0

IUPAC Name: (1S,3E)-3-{2-[(1R,3aS,4E,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
Synonyms: cholecalciferol provitina delta-D cholecalciferol [usan:ban:jan] micro-dee provitamine cholecalciferol, D3 colecalciferol
Molecular Weight (g/mol): 384.65
Molecular Formula: C27H44O
InChi Key: QYSXJUFSXHHAJI-FVUVGDFOSA-N
SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results