missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Reagent Reservoirs

Unique trough-within-trough design maximizes reagent recovery

Supplier:  VistaLab™ Technologies 30542001

 View more versions of this product

Catalog No. 21-385-101


Only null left

Description

Description

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile

Order Info

Reservoir lid sold separately (Cat. No. 21-385-110).

Specifications

Specifications

Pyrogen-free
100 mL
Sterile
General Liquid Handling
16 in.
100/Cs.
Not Autoclavable
Single-use
White
Polystyrene
Reagent Reservoir
12 in.
Individually Wrapped
14 in.
1
V-bottom
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions