missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Reagent Reservoirs
GREENER_CHOICE

Catalog No. p-165906
Change view
Click to view available options
Capacity (Metric):
100 mL (with lid)
25 mL (divided)
10 mL
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Individually packed
Individually Wrapped
Quantity:
10/bag x 10; 100/Cs.
100/Cs.
20/bag x 10; 200/Cs.
200 in. 20 bags of 10
50/Cs.
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Packaging Quantity
2138127A 100 mL Sterile Bulk 10/bag x 10; 100/Cs.
2138127B 100 mL Sterile Individually Wrapped 50/Cs.
2138127E 25 mL (divided) Sterile Bulk 10/bag x 10; 100/Cs.
21385111 100 mL (with lid) Sterile Individually Wrapped 50/Cs.
2138127D 25 mL Sterile Individually Wrapped 50/Cs.
2138127G 50 mL Sterile Bulk 10/bag x 10; 100/Cs.
2138127H 50 mL Sterile Individually Wrapped 50/Cs.
21385101 100 mL Sterile Individually Wrapped 100/Cs.
21385100 100 mL Sterile Bulk 20/bag x 10; 200/Cs.
21385104 50 mL Sterile Bulk 20/bag x 10; 200/Cs.
21385102 25 mL Sterile Bulk 20/bag x 10; 200/Cs.
21385105 50 mL Sterile Individually Wrapped 100/Cs.
21385103 25 mL Sterile Individually Wrapped 100/Cs.
22145101 25 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
22145102 25 mL (divided) Non-sterile Bulk 10/bag x 10; 100/Cs.
22145103 50 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
22145100 100 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
2138127C 25 mL Sterile Bulk 10/bag x 10; 100/Cs.
2138127F 25 mL (divided) Sterile Individually Wrapped 50/Cs.
21381090 10 mL Sterile Bulk 100/Cs.
21381091 10 mL Sterile Individually packed 50/Cs.
21381092 10 mL Sterile Bulk 200 in. 20 bags of 10
21381093 10 mL Sterile Individually packed 100/Cs.
21381094 10 mL Non-sterile Bulk 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 2138127A Supplier VistaLab™ Technologies Supplier No. 30541000
Only null left

Unique trough-within-trough design maximizes reagent recovery

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile

Order Info

Reservoir lid sold separately (Cat. No. 21-385-110).

Specifications

Certifications/Compliance Pyrogen-free
Color White
Capacity (Metric) 100 mL
Dimensions (L x W x H) 5.72 x 3.36 x 1.27 in.
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) General Liquid Handling
Height (English) 1.27 in.
Length (English) 5.72 in.
Packaging Bulk
Quantity 10/bag x 10; 100/Cs.
Width (English) 3.36 in.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less