missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Brand Precision Pipetters

Ten fixed-volume models from 10 to 1000μL

Supplier:  VistaLab™ Technologies 1112

 View more versions of this product

Catalog No. 21-381-62


Only null left
Explore available promotions

Description

Description

  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Sleeves are color coded for easy volume identification
  • Capacity engraved on bonnet

Order Info

Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.

Specifications

Specifications

±1.00%
Silver
Micropipette Tip
F1
Non-sterile
150 μL
Limited Lifetime
2 in.
Not Autoclavable
Reusable
Aluminum, Stainless Steel
1
Precision Pipette
Nozzle Insert, Calibration Key
9 in.
1.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions