Learn More
VistaLab™ Technologies MLA™ Brand Precision Pipetters
Ten fixed-volume models from 10 to 1000μL
 View more versions of this product
Catalog No. 21-338
Only null left
Explore available promotions
Description
Description
  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Sleeves are color coded for easy volume identification
  • Capacity engraved on bonnet
Order Info
Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.
Specifications
Specifications
±1.00%
Gray
Micropipette Tip
F1
Non-sterile
1000 μL
1 Ea.
9 in.
1.5 in.
Not Autoclavable
Reusable
Aluminum, Stainless Steel
1
Precision Pipette
Nozzle Insert, Calibration Key
Limited Lifetime
2 in.
Product Suggestions
Product Suggestions