missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies MLA™ Brand Precision Pipetters
Ten fixed-volume models from 10 to 1000μL
Description
- 'To deliver' capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Sleeves are color coded for easy volume identification
- Capacity engraved on bonnet
Order Info
Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.
Specifications
| ±1.00% | |
| Blue | |
| Micropipette Tip | |
| F1 | |
| Non-sterile | |
| 100 μL | |
| 1 Ea. | |
| 9 in. | |
| 1.5 in. |
| Not Autoclavable | |
| Reusable | |
| Aluminum, Stainless Steel | |
| 1 | |
| Precision Pipette | |
| Nozzle Insert, Calibration Key | |
| Limited Lifetime | |
| 2 in. |