Learn More
Applied Biosystems™ ViralSEQ™ Lentivirus Physical Titer Kit with PrepSEQ™ Nucleic Acid Sample Prep Kit, 100 reactions DFS Item
A one-step RT-qPCR assay for the quantitation of genome-containing lentiviral particles in supernatants from cell-based bioproduction systems. Lentiviral quantitation is a critical quality attribute (CQA) in viral vector manufacturing for cell and gene-modified cell therapy development.
Supplier:  Applied Biosystems™ A52598
Catalog No. A52598
Only null left
Explore available promotions
Description
Show More
Specifications
Specifications
qPCR reagents, RNA control, PrepSEQ Sample Preparation Kit, DNase I
Real-time PCR instruments (QuantStudio5 or 7500 Fast)
Lentivirus genomic target
Assay: 100 Reactions, Sample Prep: 100 Reactions
Primer-probe
100 reactions
Virus Detection
Lentivirus Physical Titer Kit, Nucleic Acid Sample Prep Kit
Product Suggestions
Product Suggestions