missing translation for 'onlineSavingsMsg'
Learn More

Violanthrone 79 95.0+%, TCI America™

Catalog No. p-7137933
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
V01311G 1 g
V01315G 5 g
2 options

Catalog No. V01311G

Supplier: TCI America V01311G

Only null left

Chemical Identifiers

CAS 85652-50-2
Molecular Formula C50H48O4
Molecular Weight (g/mol) 712.93
MDL Number MFCD00491570
InChI Key LLPQZABTDLOYAL-UHFFFAOYSA-N
Synonym violanthrone-79, violanthrone 79, anthra 9,1,2-cde benzo rst pentaphene-5,10-dione,16,17-bis octyloxy, anthra 9,1,2-cde benzo rst pentaphene-5,10-dione,1, 16,17-bis n-octyloxy anthra 9,1,2-cde benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy-anthra 9,1,2-cde benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy anthra 9,1,2-cde-benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy anthra 9,1,2-cde benzo rst pentaphene-5,10-dione
PubChem CID 16217791
SMILES CCCCCCCCOC1=C2C3=C(C=CC4=C3C(=C1)C5=CC=CC=C5C4=O)C6=C7C2=C(C=C8C7=C(C=C6)C(=O)C9=CC=CC=C98)OCCCCCCCC

Specifications

Melting Point 199°C
Quantity 1 g
Formula Weight 712.93
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Violanthrone 79