missing translation for 'onlineSavingsMsg'
Learn More

Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™

Catalog No. p-2799251
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209391000 100 mL AcroSeal™ Glass bottle
AC209398000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC209391000 Supplier Thermo Scientific Chemicals Supplier No. 209391000
Only null left

CAS: 1826-67-1 | C2H3BrMg | 131.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1826-67-1 , 109-99-9
Molecular Formula C2H3BrMg
Molecular Weight (g/mol) 131.26
MDL Number MFCD00000042
InChI Key XHHHAXOHMKAOSL-UHFFFAOYSA-M
Synonym vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide, grignard reagent
PubChem CID 74584
SMILES Br[Mg]C=C

Specifications

Color Brown
Linear Formula H2C=CHMgBr
Concentration 0.7 to 0.9M
Density 0.9800g/mL
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name bromo(ethenyl)magnesium
Formula Weight 131.25
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.98
Quantity 100 mL
Flash Point −17°C
Physical Form Solution
Chemical Name or Material Vinylmagnesium bromide, 0.7M Solution in THF
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement

Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
Precautionary Statement GHS P Statement
Store in a dry place. Store in a closed container.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only