missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinyl Palmitate (stabilized with MEHQ) 96.0+%, TCI America™
Supplier: TCI America P0848500ML
Specifications
| Vinyl Palmitate (stabilized with MEHQ) | |
| White-Yellow | |
| MFCD00059288 | |
| Palmitic Acid Vinyl Ester | |
| CCCCCCCCCCCCCCCC(=O)OC=C | |
| 282.468 | |
| 282.47 | |
| Crystalline Lumps |
| 693-38-9 | |
| C18H34O2 | |
| 500 mL | |
| UJRIYYLGNDXVTA-UHFFFAOYSA-N | |
| ethenyl hexadecanoate | |
| 69658 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 693-38-9 | |
| 282.468 | |
| UJRIYYLGNDXVTA-UHFFFAOYSA-N | |
| 69658 | |
| CCCCCCCCCCCCCCCC(=O)OC=C |
| C18H34O2 | |
| MFCD00059288 | |
| Palmitic Acid Vinyl Ester | |
| ethenyl hexadecanoate |
Safety and Handling
EINECSNumber : (2)-4160
TSCA : Yes
Recommended Storage : Refrigerator