missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinyl Palmitate (stabilized with MEHQ) 96.0+%, TCI America™
$575.62 - $575.62
Chemical Identifiers
| CAS | 693-38-9 |
|---|---|
| Molecular Formula | C18H34O2 |
| Molecular Weight (g/mol) | 282.468 |
| MDL Number | MFCD00059288 |
| InChI Key | UJRIYYLGNDXVTA-UHFFFAOYSA-N |
| Synonym | Palmitic Acid Vinyl Ester |
| PubChem CID | 69658 |
| IUPAC Name | ethenyl hexadecanoate |
| SMILES | CCCCCCCCCCCCCCCC(=O)OC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P0848500ML
|
TCI America
P0848500ML |
500 mL |
Each for $575.62
|
|
|||||
Chemical Identifiers
| 693-38-9 | |
| 282.468 | |
| UJRIYYLGNDXVTA-UHFFFAOYSA-N | |
| 69658 | |
| CCCCCCCCCCCCCCCC(=O)OC=C |
| C18H34O2 | |
| MFCD00059288 | |
| Palmitic Acid Vinyl Ester | |
| ethenyl hexadecanoate |
Specifications
| 693-38-9 | |
| C18H34O2 | |
| 500 mL | |
| UJRIYYLGNDXVTA-UHFFFAOYSA-N | |
| ethenyl hexadecanoate | |
| 69658 | |
| ≥96.0% (GC) | |
| Vinyl Palmitate (stabilized with MEHQ) |
| White-Yellow | |
| MFCD00059288 | |
| Palmitic Acid Vinyl Ester | |
| CCCCCCCCCCCCCCCC(=O)OC=C | |
| 282.468 | |
| 282.47 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-4160
TSCA : Yes
Recommended Storage : Refrigerator