missing translation for 'onlineSavingsMsg'
Learn More

Vinyl chloride Solution, 200 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020672
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020672 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020672 Supplier Merck Emd Millipore Supplier No. SGMA48625

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cleaning Products, Cosmetics, Environmental, Food and Beverages, Personal Care and Petroleum

Specifications

Packaging Ampule
CAS 75-01-4
Flash Point 11°C
Type Food and Cosmetic Component Standard
Quantity 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 200 μg/mL in methanol