missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Vinpocetine, 98%
Specific calcium-calmodulin-dependent phosphodiesterase inhibitor
$212.97 - $715.86
Chemical Identifiers
| CAS | 42971-09-5 |
|---|---|
| Molecular Formula | C22H26N2O2 |
| Synonym | Eburnamenine-14-carboxylic acid ethyl ester |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ61000FPL
|
Thermo Scientific Chemicals
J61000FPL |
20 mg |
Each for $212.97
|
|
|||||
|
AAJ61000MC
|
Thermo Scientific Chemicals
J61000MC |
100 mg |
Each for $715.86
|
|
|||||
Description
Specific calcium-calmodulin-dependent phosphodiesterase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 42971-09-5 | |
| C22H26N2O2 | |
| Eburnamenine-14-carboxylic acid ethyl ester |
Specifications
| White | |
| 42971-09-5 | |
| 350.46g/mol | |
| 147°C to 149°C | |
| Powder | |
| CCOC(=O)C1=C[C@]2(CC)CCCN3CCC4=C([C@H]23)N1C1=CC=CC=C41 | |
| 350.46 |
| Specific calcium-calmodulin-dependent phosphodiesterase inhibitor | |
| C22H26N2O2 | |
| Eburnamenine-14-carboxylic acid ethyl ester | |
| Signal Transduction Reagent | |
| DDNCQMVWWZOMLN-IRLDBZIGSA-N | |
| ethyl (15S,19S)-15-ethyl-1,11-diazapentacyclo[9.6.2.0²,⁷.0⁸,¹⁸.0¹⁵,¹⁹]nonadeca-2,4,6,8(18),16-pentaene-17-carboxylate | |
| 98% |
Safety and Handling
EINECSNumber : 256-028-0
Recommended Storage : Ambient temperatures
RUO – Research Use Only