missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinpocetine, 98%

Catalog No. AAJ61000FPL
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61000FPL 20 mg
AAJ61000MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61000FPL Supplier Thermo Scientific Chemicals Supplier No. J61000FPL

Specific calcium-calmodulin-dependent phosphodiesterase inhibitor

Specific calcium-calmodulin-dependent phosphodiesterase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42971-09-5
Molecular Formula C22H26N2O2
Synonym Eburnamenine-14-carboxylic acid ethyl ester

Specifications

Quantity 20 mg
Description White
For Use With (Application) Specific calcium-calmodulin-dependent phosphodiesterase inhibitor
Formula Weight 350.46g/mol
Melting Point 147°C to 149°C
Product Type Signal Transduction Reagent
Form Powder
InChI Key DDNCQMVWWZOMLN-IRLDBZIGSA-N
SMILES CCOC(=O)C1=C[C@]2(CC)CCCN3CCC4=C([C@H]23)N1C1=CC=CC=C41
IUPAC Name ethyl (15S,19S)-15-ethyl-1,11-diazapentacyclo[9.6.2.0²,⁷.0⁸,¹⁸.0¹⁵,¹⁹]nonadeca-2,4,6,8(18),16-pentaene-17-carboxylate
Molecular Weight (g/mol) 350.46
Assay 98%
Show More Show Less
EINECSNumber 256-028-0
Recommended Storage Ambient temperatures

RUO – Research Use Only