missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Vinpocetine, 98%
Description
Specific calcium-calmodulin-dependent phosphodiesterase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 mg |
| Description | White |
| For Use With (Application) | Specific calcium-calmodulin-dependent phosphodiesterase inhibitor |
| Formula Weight | 350.46g/mol |
| Melting Point | 147°C to 149°C |
| Product Type | Signal Transduction Reagent |
| Form | Powder |
| Assay | 98% |
RUO – Research Use Only