missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Flexible Paddle Scraper

Flexible paddle scraper is used to remove remaining food stuff from containers prior to the cleaning procedure.

$29.12 - $33.27

Specifications

Product Type Scraper
Color-coded Yes
Certifications/Compliance FDA Compliant
Description Thread type Euro Hygienic
Dimensions (L x W) 8.75 x 4.3 in.
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
17227747
View Documents Promotion Details
Vikan™
REM70134
Red
Each for $29.12
Only null left
 
29635598
View Documents Promotion Details
Vikan™
7013 5
White
Each for $33.27
Only null left
 
Description

Description

  • Intended for scraping substances from the sides of containers
  • Ideal tool for prepping prior to the cleaning procedure
  • Euro-thread connection allows for pairing with any length or type of Vikan™ handle
  • Can be used on everything from large vats to simple mixing bowls
  • Color-coded
Specifications

Specifications

Scraper
FDA Compliant
8.75 x 4.3 in.
1 Ea.
0.26 lb.
Yes
Thread type Euro Hygienic
Polyethylene
Medium
Videos
SDS
Documents

Documents