missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Single Blade Ultra Hygiene Bench Squeegee, 16 in.

Squeegee is ideal for removing water or food debris from food contact surfaces.

Specifications

Product Type Single-blade Squeegee
Certifications/Compliance FDA Compliant
Description Thread type Euro Hygienic
Includes Thread type Euro Hygienic
Material Polypropylene, Rubber
View More Specs

Products 9
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
17227835
View Documents Promotion Details
Vikan™
71404
Red N/A
Only null left
 
17227838
View Documents Promotion Details
Vikan™
71407
Orange N/A
Only null left
 
17227837
View Documents Promotion Details
Vikan™
71406
Yellow N/A
Only null left
 
17227833
View Documents Promotion Details
Vikan™
71402
Green N/A
Only null left
 
17227834
View Documents Promotion Details
Vikan™
71403
Blue N/A
Only null left
 
17227839
View Documents Promotion Details
Vikan™
71408
Purple N/A
Only null left
 
17227832
View Documents Promotion Details
Vikan™
71401
Pink N/A
Only null left
 
17227836
View Documents Promotion Details
Vikan™
71405
White N/A
Only null left
 
17227840
View Documents Promotion Details
Vikan™
71409
Black N/A
Only null left
 
Description

Description

  • One-piece construction
  • Single-blade squeegee
  • Ultra-hygienic design that minimizes areas for water and food soils to collect
  • Effective in areas requiring the highest level of hygiene
  • The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces
  • Hygiene squeegees are a good foundation for any HACCP or food safety program
  • A splash guard helps prevent water from reaching the dried surface
  • Color-coded
Specifications

Specifications

Single-blade Squeegee
Thread type Euro Hygienic
Polypropylene, Rubber
Large
FDA Compliant
Thread type Euro Hygienic
1 Ea.
3.5 in.
Videos
SDS
Documents

Documents