missing translation for 'onlineSavingsMsg'
Learn More

Vidarabine Monohydrate 98.0+%, TCI America™

Catalog No. V00981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00981G 1 g
V00985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00981G Supplier TCI America Supplier No. V00981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24356-66-9
Molecular Formula C10H15N5O5
Molecular Weight (g/mol) 285.26
MDL Number MFCD00065471
InChI Key ZTHWFVSEMLMLKT-CAMOTBBTSA-N
Synonym Ara-A, Adenine 9-beta-D-Arabinofuranoside, 9-(beta-D-Arabinofuranosyl)adenine
PubChem CID 32326
ChEBI CHEBI:9978
IUPAC Name (2R,3S,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol;hydrate
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)CO)O)O.O

Specifications

Melting Point 253°C
Color White-Yellow
Quantity 1 g
Formula Weight 267.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Vidarabine Monohydrate
EINECSNumber (5)-1138
RTECSNumber AU6200000
TSCA No