missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control - VIC™ Assay

Catalog No. A29765
Encompass
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A29765 100 Reactions
A29767 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29765 Supplier Applied Biosystems™ Supplier No. A29765

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The VetMAX™ Xeno™ Internal Positive Control (IPC) - VIC™ Assay is a primer-probe mix that detects the Xeno internal positive control. The resultant Xeno data is used to determine the validity of diagnostic test results.

The Xeno IPC assay is introduced during the qPCR preparation step and carried through the animal health PCR workflow.

Features of the VetMAX Xeno IPC - VIC Assay include:

  • Provides confidence that qPCR test results are accurate and actionable
  • Easily integrates into any workflow
  • Greatly reduces the likelihood of false negatives

The VetMAX Xeno IPC - VIC Assay comes in a 25X concentration and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with VetMAX™ Xeno Internal Positive Control RNA or VetMAX™ Xeno Internal Positive Control DNA, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC assays are included in most VetMAX kits and have been successfully benchmarked against millions of genomes including those relevant to animal health. A VetMAX™ Xeno™ Internal Positive Control - LIZ™ Assay (Cat. No. A29766) is also available.

Specifications

Concentration 25X
Content And Storage 1 x 110 μL Xeno VIC Primer Probe Mix. Store at -5°C to -30°C.
Format Kit
GC-Rich PCR Performance Low
PCR Method qPCR
Label or Dye VIC
Product Line VetMAX, Xeno
Product Type Internal Positive Control Assay
Quantity 100 reactions
Shipping Condition Dry Ice
Species Not Species-Specific
Fidelity (vs. Taq) 25 X
No. of Reactions 100 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.