missing translation for 'onlineSavingsMsg'
Learn More

VetMAX™ PRRSV NA & EU Reagents

Catalog No. 361055899
Catalog No. 361055899 Supplier Thermo Fisher Scientific Supplier No. 4468465

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VetMAX™ NA and EU PRRSV Reagents contains primers/probes that enable multiplex, one-step, reverse transcription and real-time PCR (RT-PCR) targeting North American and European Porcine Reproductive and Respiratory Syndrome Virus (PRSSV) RNA and Xeno™ RNA Control.

Note: The name of this product has changed. It was previously called PRRSV REAGENTS W/PATCH EACH.

VetMAX™ NA and EU PRRSV Reagents contains primers/probes that enable multiplex, one-step, reverse transcription and real-time PCR (RT-PCR) targeting North American and European Porcine Reproductive and Respiratory Syndrome Virus (PRSSV) RNA and Xeno™ RNA Control. VetMAX™ NA and EU PRRSV Reagents contains an updated primer/probe mix compared to the similar TaqMan™ NA and EU PRRSV Reagents.

  • Rapid and cost-effective method for amplification of target sequences
  • Quality reagents, manufactured under ISO 9001, providing confidence in the supply and quality of products
  • Over 20 years in the DNA/RNA arena, with extensive bioinformatics capabilities and experience

PRSSV causes significant losses in the swine industry due to reproductive disorders, respiratory conditions, and growth retardation. PRRSV can be detected in blood and semen 24–48 hours after infection. Real-time PCR is the most sensitive method for monitoring RNA amplification.

Specifications

Content And Storage The kit contains Multiplex RT-PCR Buffer, Multiplex Enzyme Mix, PRRSV Primer Probe Mix v2, and nuclease-free water. Store the components at -5 to -30°C, 4°C, or room temperature as indicated.
Detection Method Primer-probe
PCR Method 1-step RT-qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 Fast System, 7500 System
Passive Reference Dye ROX (Pre-mixed)
Product Line VetMAX
Product Type PCR Reagents and Detection Kit for Animal Health
Quantity 100 reactions
Reverse Transcriptase ArrayScript™ UP
Shipping Condition Dry Ice
No. of Reactions 100 Reactions
Sample Type Viral Samples
Show More Show Less