missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ PRRSV NA & EU Controls

Catalog No. 501147904
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501147904 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501147904 Supplier Applied Biosystems™ Supplier No. 4405548

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VetMAX™ NA and EU PRRSV Controls are used in multiplex, real-time reverse transcription PCR (RT-PCR) to simultaneously detect and subtype both North American (NA) and European (EU) Porcine Reproductive and Respiratory Syndrome Virus (PRRSV).

Note: The name of this product has changed. It was previously called TAQMAN PRRSV AND XENO RNA CTRL EACH.

VetMAX™ NA and EU PRRSV Controls are used in multiplex, real-time reverse transcription PCR (RT-PCR) to simultaneously detect and subtype both North American (NA) and European (EU) Porcine Reproductive and Respiratory Syndrome Virus (PRRSV). XenoRNA™ RNA Control is an internal positive control added to the lysis solution during RNA isolation to distinguish PCR inhibition, reaction failure, or human error from true negatives. In a single-tube, one-step reaction, PRRSV RNA is reverse transcribed into cDNA.

Specifications

Content And Storage Control Template(s)
Format Tube
Label or Dye Quasar 670
Quantity 1 kit
Shipping Condition Dry Ice
Species Swine/Porcine
Target Organism Class Porcine Reproductive and Respiratory Syndrome Virus (PRRSV)
For Use With (Application) Animal Health

For Research Use Only. Not for use in diagnostic procedures.