missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ PEDV/TGEV/SDCoV Kit

Catalog No. A33402
Encompass
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33402 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A33402 Supplier Applied Biosystems™ Supplier No. A33402
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A single-well multiplex real-time RT-PCR kit for the detection and differentiation of the porcine epidemic diarrhea virus (PEDV), transmissible gastroenteritis virus (TGEV), and swine delta corona virus (SDCoV) in environmental samples.

The Applied Biosystems VetMAX PEDV/TGEV/SDCoV Kit is a single-well multiplex real-time RT-PCR kit for the identification of the porcine epidemic diarrhea virus (PEDV), transmissible gastroenteritis virus (TGEV), and swine delta corona virus (SDCoV) in environmental and feed samples.

  • 4 separate tests are run per sample in 1 well—no need to run several PCRs per virus
  • The amplification can be run in standard and fast mode, depending on your needs and laboratory workflow
  • Instructions included

The kit includes:

  • TaqMan Fast Virus 1-Step Master Mix
  • VetMAX PEDV/TGEV/SDCoV Primer Probe Mix—for optimized multiplex real-time RT-PCR amplification of PEDV/TGEV/SDCoV RNA targets
  • VetMAX PEDV/TGEV/SDCoV/Xeno Control RNA Mix
  • VetMAX Xeno Positive Control RNA—serves as an internal positive control for the RNA purification process and a monitor for the presence of PCR inhibitors

Ease of use

All components for running the PCR are included in the kit. The sample preparation workflow has been tested with Applied Biosystems MagMAX sample extraction chemistry and Thermo Fisher KingFisher instruments. The kit has been validated for several sample types, eliminating the need for separate kits for each type. And this real-time PCR kit can be used with different thermocyclers, with the same results and reliability.

Contact us for more information on how to collect environmental samples on the farm or in transport vehicles.

Environmental samples are:

  • Feed samples
  • Fecal samples collected from floors or walls
  • Swiffer pads

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Primer-probe
GC-Rich PCR Performance Low
PCR Method 1-step RT-qPCR
Reaction Speed Fast or Standard
No. of Tests 100 Tests
Product Line VetMAX
Quantity 100 Tests
Shipping Condition Dry Ice
Sample Type Swiffer pads and boots, Food & Environmental Samples
What is the master mix used in the VetMAX PED/TGEV/SDCoV Kit?

The master mix used in this kit is the TaqMan Fast Virus 1-Step Master Mix.

What are the probes included in the VetMAX PEDV/TGEV/SDCoV Kit?

The TaqMan assay for PEDV RNA is labeled with LIZ, the TaqMan assay for TGEV is labeled with FAM, the TaqMan assay for SDCoV is labeled with VIC, and the TaqMan assay for the Xeno RNA is labeled with NED. All four TaqMan assays are combined in the VetMAX PEDV/TGEV/SDCoV Primer Probe Mix.

What is the master mix used in the VetMAX PEDV/TGEV/SDCoV Kit?

The master mix used in the VetMAX PEDV/TGEV/SDCoV Kit is the TaqMan Fast Virus 1-Step Master Mix.