missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ VetMAX™ M. hyopneumoniae Reagents, Applied Biosystems™ design
Description
VetMAX™ M. hyopneumoniae Reagents Applied Biosystems Design is a TaqMan™ primer probe mix for real-time PCR amplification of Mycoplasma hyopneumoniae DNA and Xeno™ DNA Internal Positive Control.
Specifications
Specifications
| Content And Storage | Store at -20°C. Includes Mhyo AB Design TaqMan™ Primer Probe mix. |
| Product Line | VetMAX |
| Product Type | PCR Reagents and Detection Kit for Animal Health |
| Quantity | 100 reactions |
| Shipping Condition | Dry Ice |
| Species | Swine/Porcine |
| Sufficient For | 100 Reactions |
| Target Organism Class | M. hyopneumoniae |
| No. of Reactions | 100 Reactions |
For Research Use Only. Not for use in diagnostic procedures.