missing translation for 'onlineSavingsMsg'
Learn More

VetMAX™ M. hyopneumoniae Controls (positive internal DNA control)

Catalog No. 501147915
Change view
Click to view available options
Quantity:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501147915 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501147915 Supplier Thermo Fisher Scientific Supplier No. 4415198

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VetMAX™ M. hyopneumoniae Controls is a mixture of synthetic Mycoplasma hyopneumoniae DNA and Xeno™ DNA Internal Positive Control, used as a positive control mix for real-time PCR amplification of M. hyopneumoniae DNA.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Content And Storage Store at -20°C. Includes Mhyo-Xeno™ DNA Control Mix.
Form Liquid
Format Tube
Reaction Speed Standard
Product Line VetMAX
Product Type PCR Reagents and Detection Kit for Animal Health
Quantity 10 reactions
Shipping Condition Dry Ice
Species Swine/Porcine
Sufficient For 10 Reactions
Target Organism Class M. hyopneumoniae
No. of Reactions 10 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.