missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VetMAX™-Gold SIV Subtyping Kit
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Includes: 2X Multiplex RT-PCR buffer 2X Multiplex enzyme mix 2X Nuclease-free water 2X SIV N1/N2 primer-probe mix 2X SIV H1/H3 primer-probe mix 2X SIV Typing RNA control mix Store at -5 to -30°C. |
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | Low |
| PCR Method | 1-step RT-qPCR |
| No. of Tests | 100 tests |
| Product Line | VetMAX |
| Product Type | Real Time PCR Reagents |
| Quantity | 100 tests |
| Fidelity (vs. Taq) | 2 X |
For Veterinary Use Only