missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VetMAX™-Gold BVDV PI Detection Kit
Specifications
Specifications
| Content And Storage | Store at -30°C to -10°C. Includes: RT-PCR Enzyme Mix RT-PCR Buffer BVDV Primer Probe Mix Xeno™ RNA Control BVDV Control RNA Nucleic Acid Dilution Solution Store at -30°C to 25°C: Nuclease-free Water |
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | DNA Polymerase |
| Reaction Speed | Fast |
| Product Line | VetMAX |
| Product Type | PCR Reagents and Detection Kit for Animal Health |
| Quantity | 100 reactions |
| Show More |
Frequently Asked Questions (FAQs)
With the VetMAX-Gold BVDV PI Detection Kit, I am getting signal detection in the no-template control (NTC) reaction. Why is this?
This is likely due to PCR contamination. Here are some recommendations:
- Repeat the qRT-PCR reaction with fresh reagents and freshly decontaminated pipettors.
- Set up and run the qRT-PCR reaction in an area that is isolated from areas used for RNA purification and PCR product analysis.
For Veterinary Use Only