missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Verso cDNA Synthesis Kit

Catalog No. p-4526924
Change view
Click to view available options
No. of Reactions:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
AB1453B 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1453B Supplier Thermo Scientific™ Supplier No. AB1453B
Only null left

The Thermo Scientific Verso cDNA Synthesis Kit provides robust transcription of RNA to create a complete cDNA pool.

The Thermo Scientific Verso cDNA Synthesis Kit provides robust transcription of RNA to create a complete cDNA pool. The Verso system achieves robust and sensitive reverse transcription through the combination of a high affinity RT enzyme, a unique RNA priming method, and an optimized buffering system. The Verso RT enzyme has high RNA template affinity and reduced RNase H activity, to transcribe even sections with high secondary structure. The included anchored oligo dT priming method further enhances sensitivity by increasing transcription efficiency, and the cDNA synthesis buffer has been optimized to achieve a full and diverse cDNA pool.

Features of the Verso cDNA Synthesis Kit include:
• Highly sensitive with a broad dynamic range—can reverse transcribe RNA from an input range of 1pg to 1μg
• Efficient RT reaction allows for 75% shorter protocol times (down to 30 minutes)
• Unique priming strategy for increased efficiency and broad template flexibility
• High affinity RT enzyme effectively transcribes through difficult RNA sequences
• Wide working temperature range (42–57°C) for success with GC-rich and other difficult templates
• Included RT enhancer prevents genomic DNA carryover, eliminating the need for separate DNase I treatment
• RNase inhibitor included
• Can be paired with any PCR Kit for complete 2-Step RT-(q)PCR

Applications
• 2-step RT-(q)PCR
• Single stranded cDNA synthesis

Specifications

Content And Storage

• RT Enzyme mix with RNase Inhibitor
• 5X RT Buffer
• Random Hexamers
• Anchored Oligo (dT)
• 20 mM dNTPs
• RT Enhancer

Store at –20°C.

Format Kit
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Quantity 100 reactions
Reverse Transcriptase Verso
Ribonuclease H Activity Yes
Shipping Condition Wet or Dry Ice
For Use With (Application) Real Time PCR (qPCR), RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 100 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 11 kb
Starting Material RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.