Learn More
Thermo Scientific™ Verso 1-step RT-qPCR Kit, SYBR Green, low ROX
Description
The Thermo Scientific Verso 1-step RT-qPCR Low ROX Kit for SYBR Green chemistry offers a quick and simple way to detect mRNA.
An optional RT Enhancer is also provided and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment. All Verso 1-step RT-qPCR kits incorporate an inert dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.
In a 1-step approach to RT-qPCR, both the RT step and qPCR steps are performed in a single tube (1-step RT-qPCR). Adopting a 1-step approach is ideal for primer sequence optimization and collecting comparative data in experiments that do not require subsequent cDNA storage. The approach minimizes the risk of contamination, and improvements in reproducibility may be observed as a result of reduced sample handling.
Verso One-Step RT-qPCR Kits for SYBR Green chemistry are compatible with all major real-time thermal cyclers.
- Eliminate the need for DNase treatments - RT Enhancer (optional) prevents gDNA carry-over
- RT and qPCR steps performed in single tube - convenience and reduced contamination risk
- Use of the entire RT reaction as template for qPCR - improved sensitivity
- Verso RT enzyme is highly sensitive with broad dynamic range - accurate detection of RNA from 1pg to 1μg
- Verso RT enzyme has a wide working temperature range (42° to 57°C) - improved success with GC-rich and other difficult templates
- Contains an inert blue dye for ease of pipetting during reaction setup
Specifications
Specifications
| Content And Storage | Contains: • 1 x 50 μL Verso Enzyme Mix • 1 x 250 μL RT enhancer • 2 x 1.25 mL 2X 1-Step qPCR SYBR Low ROX Mix • 1 x 100μL 1M MgCl2. Sufficient for 200 x 25 μL reactions. Store at -20°C |
| Detection Method | SYBR |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | 1-step RT-qPCR |
| For Use With (Equipment) | 7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P |
| Product Line | Verso |
| Product Type | qPCR Mix |
| Quantity | 200 reactions |
| Sufficient For | 200 Reactions |
| Show More |
Frequently Asked Questions (FAQs)
The RT Enhancer is a unique enzyme that degrades contaminating genomic DNA at the start of the RT step. It is equally as effective as DNase I treatment but more convenient as it eliminates the need for a separate DNase I incubation prior to RT-PCR.
No, you do not need to treat your RNA with DNase I before running RT-qPCR with the Verso cDNA Synthesis Kit. The RT enhancer included in the Verso kit degrades any contaminating double-stranded DNA during the reverse transcription process, eliminating the need for a separate DNase I treatment.
No, we do not offer the Verso Enzyme Mix as a standalone product. It is only available as a component of the Verso 1-step RT-qPCR Kit (Cat. No. AB4016A, AB4016C).
For Research Use Only. Not for use in diagnostic procedures.