missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Yttrium (Y) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$252.09 - $547.44
Chemical Identifiers
| CAS | 13494-98-9 |
|---|---|
| Molecular Formula | H12N3O15Y |
| Molecular Weight (g/mol) | 383.008 |
| InChI Key | QBAZWXKSCUESGU-UHFFFAOYSA-N |
| PubChem CID | 166833 |
| IUPAC Name | yttrium(3+);trinitrate;hexahydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010060100
|
Ricca Chemical Company
RV010060100N |
100 mL |
Each for $252.09
|
|
|||||
|
RV010060500
|
Ricca Chemical Company
RV010060500N |
500 mL |
Each for $547.44
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 13494-98-9 | |
| 383.008 | |
| 166833 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] |
| H12N3O15Y | |
| QBAZWXKSCUESGU-UHFFFAOYSA-N | |
| yttrium(3+);trinitrate;hexahydrate |
Specifications
| Natural LDPE Bottle | |
| 95.62%,1.96%,0.42% | |
| 1.01 | |
| H12N3O15Y | |
| QBAZWXKSCUESGU-UHFFFAOYSA-N | |
| yttrium(3+);trinitrate;hexahydrate | |
| 100 mL | |
| High Purity |
| 13494-98-9,7697-37-2,7732-18-5 | |
| 99.52%,2.04%,0.44% | |
| Liquid | |
| Approximately 0°C | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] | |
| 383.008 | |
| 166833 |