missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Trace Metals Standard I, Ricca Chemical

Catalog No. RV010852100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010852100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010852100 Supplier Ricca Chemical Company Supplier No. RV010852100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

10 ppm Hg, 25 ppm Cd, Se; 100 ppm: As, Be, Co, Cr, Cu, Fe, Mn, Ni, Pb, Zn; 250 ppm V, 500 ppm Al /nM

  • 10ppm Hg, 25ppm Cd, Se; 100ppm: As, Be, Co, Cr, Cu, Fe, Mn, Ni, Pb, Zn; 250ppm V, 500ppm Al
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 4.90%
CAS Max % 5.10%
Specific Gravity 1.08
Physical Form Liquid
UN Number UN3264
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Multi-element Standard Solution
Quantity 100 mL
Grade High Purity
Chemical Name or Material Trace Metals Standard
Show More Show Less