missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Titanium Standard for ICP 1000 ppm 5% HCl/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010322500N
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 92.30%,4.90%,0.49% | |
| 1 | |
| FH | |
| UN3264 | |
| Standard | |
| F | |
| 20.01 | |
| 14917 | |
| High Purity |
| 7664-39-3 , 7647-01-0 , 7664-39-3 | |
| 96.06%,5.10%,0.51% | |
| Liquid | |
| MFCD00011346 | |
| Approximately 0°C | |
| KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
| hydrogen fluoride | |
| 500 mL | |
| CHEBI:29228 |
Chemical Identifiers
| 7664-39-3 | |
| 20.01 | |
| KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
| CHEBI:29228 | |
| F |
| FH | |
| MFCD00011346 | |
| 14917 | |
| hydrogen fluoride |